GENPack for KidWind
Supplier: Vernier Software & Technology
KW-GP
470165-144EA 78.99 USD
470165-144
GENPack for KidWind
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
The GENPack allows students to construct their own electrical generator and perform experiments with electricity and magnetism. Changing variables in the generator design will impact current and voltage output. The GENPack is perfect for physics and electrical students exploring Faraday’s law, induction, AC/DC electricity production, and energy transformations. We recommend this kit for advanced middle school students and older.