Polyproplyene Test Tube Drying Racks
Supplier: United Scientific Supplies
77852
470164-058PK 66.9 USD
470164-058
Polyproplyene Test Tube Drying Racks
Racks Draining Racks
Dry up to 50 tubes at once.
  • Autoclavable
  • 5×10 array of pegs
  • Conveniently sold in package of 6 racks
These polypropylene racks can conveniently hold inverted tubes for drying and to minimize collection of airborne contaminants. Rack can hold up to 50×16 mm diameter tubes. Racks can also be used to hold and dry electrophoresis and chromatography plates.