Geoblox Hurricane Katrina Disaster Model Set
Supplier: JOHN HAROLD KOONZ
DEFAULT VPN
470154-412EA 30.75 USD
470154-412
Geoblox Hurricane Katrina Disaster Model Set
Models Meteorology Models
Examine the Science Behind the Landmark 2005 Hurricane
An event in recent memory for your earth science students, Hurricane Katrina can teach a great deal about disaster planning, severe storms, and environmental effects. This set of 19 model patterns examines many aspects of the New Orleans flooding, including Meandering Stream, Natural Levees and Floodplain Deposition, Barrier Islands, Delta Deposition and Distributary Shifting, Subsidence Due to Natural Tectonic Activity, Artificial Levees Disrupt Deposition, Mississippi River Gulf Outlet, Wetlands Before and After Spoil Banks, OilMore Product Information