Bacterial Gram Staining Activity Kit
Supplier: G-Biosciences
BE-202
470149-972KT 185.6 USD
470149-972
Bacterial Gram Staining Activity Kit
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Demonstrate a common diagnostic technique.
  • Teach students a diagnostic test done in labs every day
  • Kit includes everything you need to stain bacteria
Teach students the basics of Gram staining procedure, a valuable diagnostic tool commonly used to identify bacteria in research and clinical settings. Students will study two different strains of bacteria under a microscope to differentiate Gram-positive and Gram-negative bacteria.
Kit includes bacteria strains and all necessary components for Gram staining.
Supplied with components needed forMore Product Information