Recombinant DNA Manipulative

Supplier: United Scientific Supplies
DNAKIT
470144-216EA 425.1 USD
470144-216
Recombinant DNA Manipulative
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
This large plasmid model teaches students the basics of recombinant DNA technology.

  • Students Learn the Basics of Genetic Engineering
  • Components Made of Durable Plastic
  • Magnetic Strips Connect the Plasmid Model Pieces
  • Material for 15 Demonstrations
  • Manual and Instructional Video Included

The teacher explains the process of cutting plasmid DNA with restriction enzymes while students follow along using the included manipulative sets. The teacher then uses the laminated electrophoresis boards to represent the separation of fragments on a gel.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR