Electrostatic Kit
Supplier: Wards
PH0900
470120-700EA 52.2 USD
470120-700
Electrostatic Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Detect the Presence of Static Electricity
  • User Friendly
  • Reliable Materials
Charge electroscopes, show electrostatic attraction, and more with the contents of this activity kit. A simple electroscope is included for easily identifying static electricity. A series of other materials including metalized polystyrene spheres, polyethylene tiles, aluminum cans, nylon, PVC, wire, rubber cloth, and wire hook are also supplied for completing electricity experiments.