Electrophoresis Running Buffer
Supplier: Edvotek
607
470111-864EA 41.6 USD
470111-864
Electrophoresis Running Buffer
Buffers Electrophoresis Buffers Running Buffers
Tris Acetate, pH 7.8, concentrated 50X (for 5 L—20 standard gels).