DC Motor

Supplier: Wards
PH1242
470111-202EA 29.9 USD
470111-202
DC Motor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This DC Motor arrives completely assembled.

  • Storage Case Included
  • Open Design
  • Reversing Magnets Changes the Direction of Motor's Rotation
  • Motor Can Be Used Vertically or Horizontally
  • For Experiments and Demonstrations

This open design is similar to diagrams in most textbooks and makes it easy to study motor-related principles. Users can reverse magnets to change direction of the motor’s rotation. Users can also observe change of speed with current with an external rheostat (not included).

The lightweight motor can be used vertically or horizontallyMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR