Riker Insect Mounts
Supplier: Jordan Paper Box
EX8X12PB
470104-312CS 450.89 USD
470104-312
Riker Insect Mounts
Jars Specimen Jars
Keep your specimens flat and protected with these quality insect display mounts.
These cases feature a clear glass front and a cotton lining designed to preserve your samples. Your students can use these mounts to show the life histories and natural surroundings of various insect and plant specimens. In addition, a complete specimen record can be written on the back. Each mount is approximately 5 cm deep.