Ward's® Tryptic Soy Broth Dehydrated Media, Powdered, IPM Scientific
Supplier: IPM SCIENTIFIC, INC.
M011-500G
470024-770EA 72.55 USD
470024-770
Ward's® Tryptic Soy Broth Dehydrated Media, Powdered, IPM Scientific
Media for Microbiology Dehydrated Media for Microbiology
Dilute quantities to any volume as needed.
  • Economical alternative
  • Stores much longer than prepared media
  • Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.