Blood Based Cancer Diagnosis Lab Activity
Supplier: Edvotek
141
470024-510EA 139 USD
470024-510
Blood Based Cancer Diagnosis Lab Activity
Educational Classroom Kits and Activities
Cancer cells differ from normal cells by the combination of normal proteins that are present on their surfaces.
  • Students Observe the Reaction of Cancer Cell Markers and Their Corresponding Antigens
  • Instructions Provided
  • Lab Takes 35 Minutes to Complete
  • Material for 10 Student Groups Provided
Antibodies against these proteins specifically bind to cancer cells but not normal cells. This characteristic allows for early detection of cancer and potentially a way of delivering cancer therapies.
In this experiment, students view this process first handMore Product Information