Beaker Tongs
Supplier: United Scientific Supplies
BTSP09
470019-542EA 22.3 USD
470019-542
Beaker Tongs
Tongs
Equipped with convenient plastic coated jaws.
  • Zinc-plated steel
  • Opens to 7"
  • Overall length is 9.75"
These beaker tongs are constructed with flat riveted joints and convenient vinyl coated sleeves. Tongs can handle beakers ranging from 50 to 1500 mL capacity.