Wire Gauzes, Plain
Supplier: United Scientific Supplies
WGPL44-PK/12 WGPL55-PK/12 WGPL66-PK/12 WGPL66 WGPL55 WGPL44 WGPL44 WGPL55 WGPL66
470019-498PK 21.15 USD
470019-498 470019-500 470019-502 470144-816 470144-818 470144-820 470176-062 470188-996 470188-998
Wire Gauzes, Plain
Chemicals for Science Education
Sturdy wire mesh for supporting glassware over a Bunsen burner.
  • Long lasting, heat- and tear-resistant iron
  • Available in a variety of sizes
Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.