Horseshoe Magnet
Supplier: United Scientific Supplies
MHS030
470018-704EA 10.15 USD
470018-704
Horseshoe Magnet
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Teeny but Strong!
  • Keepers helped retain the magnetic strength
  • Able to pull 6 pounds
This powerful permanent magnet weighs only 1 oz., but pulls 6 lbs. It comes with a soft iron “keeper” and has a gap of 3/4" at the open end.