Compound Bar
Supplier: American Scientific
3230-00
470005-408EA 16.2 USD
470005-408
Compound Bar
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Laminated for long use.
  • Bar length is 20 cm
  • Bar and handle length is 30 cm
A laminated bar of two metals fastened in a wooden handle are placed over a flame. The different thermal expansion rates cause the laminated bar to bend.