VeriClean™ Defluxer Solvent, MicroCare
Supplier: MicroCare
MCC-DC1
46620-506CS 405.39 USD
46620-506
VeriClean™ Defluxer Solvent, MicroCare
Soldering Supplies Flux Removers
Powerful, ozone- and people-safe defluxer and medium-strength degreaser based on ultra-pure siloxanes
Works well on rosin and “no-clean” fluxes and pastes, almost any oil or light grease, and many adhesives. It also melts away “solventless” silicone-based conformal coatings. Fast-drying and odorless. High purity (<2ppm NVR guaranteed) and great toxicity ratings. Plastic-safe, ESD-safe, and residue-free. Requires no rinsing. Flammable. Aerosol spray can.
Order Now
SPECIFICATIONS
Environmentally Preferable