VWR® 9 mm Screw-Thread Vials
Supplier: VWR International
66009-852 66009-856 66009-860 82030-970 82030-974 82030-976 46610-722 46610-724 46610-726 66009-854 66009-858 66009-862 89239-024 89239-026 89239-028 89239-030 89524-188 89524-190 10781-890 10803-884 10803-886 10803-888 10804-054 10804-056 76326-138 76326-140 76326-142 76326-148
66009-852CS 99.68 USD
66009-852 66009-856 66009-860 82030-970 82030-974 82030-976 46610-722 46610-724 46610-726 66009-854 66009-858 66009-862 89239-024 89239-026 89239-028 89239-030 89524-188 89524-190 10781-890 10803-884 10803-886 10803-888 10804-054 10804-056 76326-138 76326-140 76326-142 76326-148
VWR® 9 mm Screw-Thread Vials
Vials
Autosampler screw thread vials feature a wide mouth design for compatibility with all major autosampler brands and robotics.
  • Precision manufactured with high-purity borosilicate glass
  • Caps and vials matched for best closure integrity
  • Available with or without writing patch
  • Compatible with poly-spring inserts
  • Purchase vial and caps in convenience kit or separately
  • Available pre-assembled, certified, clear or amber glass
Vials are manufactured from Type 1 borosilicate glass and have a maximum fill volume of 1.8 ml. Certified vial kits are tested andMore Product Information