PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Supplier: Corning
7103C-1 7103C-2 7103C-3 7103C-4 7103C-5 7103C-6 7103C-7 7103C-8 7103C-9 7103C-10 7103C-15 7103C-20 7103C-25
46610-102CS 234.32 USD
46610-102 46610-104 46610-106 46610-108 46610-110 46610-112 46610-114 46610-116 46610-118 46610-120 46610-122 46610-124 46610-126
PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Pipettes Graduated Pipettes
These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Reusable precision pipettes
  • Color-coded markings are enameled onto the glass
PYREX® serialized/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line)More Product Information