Diluent 2 for hematology
Supplier: Thermo Fisher Scientific
DIL5522
45001-012EA 92.49 USD
45001-012
Diluent 2 for hematology
Diluent 2
Hematology reagents for use with Beckman Coulter® analyzers not Performing WBC Differentials.
  • Multi-Purpose Blood Diluent