Corning® MEM Vitamins 100X Solution, Corning
Supplier: Corning
25-020-CI
45000-702PK 178.9 USD
45000-702
Corning® MEM Vitamins 100X Solution, Corning
Cell Culture Supplements and Additives
The addition of MEM vitamins to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells
This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.