Corning® HT (Hypoxanthine, Thymidine), Corning
Supplier: Corning
25-047-CI
45000-688EA 32.77 USD
45000-688
Corning® HT (Hypoxanthine, Thymidine), Corning
Cell Culture Media
HT media is a 'rescue media' that provides pre-formed purines and pyrimidines to overcome the effects of residual intracellular aminopterin.
  • Complete Certificate of Analysis available for each production lot, along with full formulation information
  • Produced under the highest industry standards to ensure superior results