Corning® Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA, without Phenol Red, Corning
Supplier: Corning
40-102-CV
45000-422PK 243.75 USD
45000-422
Corning® Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA, without Phenol Red, Corning
Cell Culture Media
Corning® cellgro® COMPLETE formulation is based on a combination of 1:1 DMEM/F12, RPMI, and McCoy's 5A medium. This medium contains a proprietary mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1.0g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspensions and adherent cell cultures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage Temperature Refrigerate