Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Supplier: Corning
10-050-CV 10-051-CI
45000-374PK 238.03 USD
45000-374 45000-376
Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Cell Culture Media
McCoy's 5A medium was originally formulated to meet the culture requirements of Novikoff Hepatoma cells, and has been found to support the indefinite proliferation of Walker256 carcinoma cells along with a wide range of human and rat normal and transformed cells.  Originally modified by Iwakata and Grace, this version of McCoy's 5A includes increased levels of inositol and glucose
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment