NOVA-TECH™ Lint Free Nonwoven Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
NT1-77 NT1-66 NT1-44 NT10-99 NT1-1818
43500-100CS 626.44 USD
43500-100 10129-546 10129-548 10129-550 10129-572 10129-549 10129-553
NOVA-TECH™ Lint Free Nonwoven Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
NOVA-TECH™ Lint Free Wipes are a popular general purpose non-woven wipe comprised of blended cellulose and polyester. They are tough, abrasion resistant, absorbent, and virtually free of particle generation.
  • Perfect for use in class 1000 cleanroom or higher
  • Absorbent
  • Strong, ideal for general wiping
The synergistic blend of virgin polyester and cellulose is hydro-entangled into a uniform, super-tough fabric. These features give NOVA-TECH™ Lint Free Wipes a high degree of absorbency and effectiveness during the process of general wiping.More Product Information