Microfine Grinding Mill, MF 10, IKA® Works
Supplier: IKA Works
2836000 0002836001
43300-186EA 6298.19 USD
43300-186 43300-188
Microfine Grinding Mill, MF 10, IKA® Works
Mills
This continuous microfine, stainless steel grinder features interchangeable grinding heads
Choose the cutting grinding head for crushing fibrous substances such as paper and vegetation, as well as plastics and other materials with a low volume weight. The impact grinding head crushes brittle and hard materials such as minerals and building materials.
Ordering information: A grinding head and a 304 stainless steel sieve are required for operation and must be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable
Dimensions 32W x 30D x 38H cm ( 12 19/32 x 11 13/16 x 14 61/64 ) 31W x 37D x 56.9H cm (127/32 x 1419/32 x 2213/32")
Max. Working Capacity 5 kg/hr
Weight 9.7 kg (21.3 lbs.) 10 kg (22 lbs.)