VWR® CryoCooler Freeze Controller
Supplier: VWR International
414004-284
414004-284CS 738.79 USD
414004-284
VWR® CryoCooler Freeze Controller
Coolers Benchtop Coolers
When filled with isopropyl alcohol and placed in mechanical freezer, this polycarbonate freeze controller provides the -1°C per minute cooling rate required to successfully preserve cells.
  • Non-mechanical
  • Successfully preserves cells
  • Insert helps prevent contamination
Floating rigid insert helps prevent contamination. Withstands repeated use. Holds up to eighteen 1.2–2 mL cryogenic vials.