VWR® CERTICLEAN® Cleanroom Nitrile Gloves, Class 10
Supplier: VWR International
40101-357 40101-352 40101-353 40101-354 40101-356 40101-358
40101-357CS 761.1 USD
40101-357 40101-352 40101-353 40101-354 40101-356 40101-358
VWR® CERTICLEAN® Cleanroom Nitrile Gloves, Class 10
Gloves
Ambidextrous, powder-free gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Superior strength and puncture resistance
  • Certificates of analysis available
  • Quality data available
Nitrile is superior in strength and puncture resistance compared to other glove compounds. Film thickness designed for comfort and tactile sensitivity. Gloves are 6.3 mil thick at fingers and 5.1 mil thick at palms. Textured fingertips for a reliable grip. Uncompromising manufacturing processes yield consistently low particle counts. Proprietary nitrile,More Product Information