Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RTK4530]

Supplier: Biolegend
400628
400628-BLEA 248.08 USD
400628-BL
Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RTK4530]
Antibodies
Alexa Fluor® 700 Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 100 μg

This reagent is developed for immunofluorescent staining for flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. It is highly recommended that the reagent be titrated for optimal performance for each application.

* Alexa Fluor® 700 has a maximum emission of 719 nm when it is excited at 633nm / 635nm. Prior to using Alexa Fluor® 700 conjugate for flow cytometric analysis, please verify your flowMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol IgG2b k isotype ctrl
Environmentally Preferable
Antibody Type Control
Clonality Monoclonal
Conjugation Alexa Fluor® 700
Clone RTK4530
Reactivity Rat
Host Rat
Isotype IgG2b kappa
Flow Cytometry Yes
Immunogen Trinitrophenol + KLH
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The immunoglobulin was purified by affinity chromatography, and conjugated with Alexa Fluor® 700 under optimal conditions.
Concentration 0.50 mg/mL
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR