KV1000 Digital Constant Temperature Kinematic Viscosity Bath, 220-240 V, Koehler
Supplier: Koehler
K23377-00000 K23378-00000
37000-160EA 12736.57 USD
37000-160 37000-164
KV1000 Digital Constant Temperature Kinematic Viscosity Bath, 220-240 V, Koehler
Baths Viscometer Baths
Single phase, constant temperature bath for kinematic viscosity testing of petroleum products
  • 220-240 V models, not for use in USA
Accommodates six round 51mm (2") diameter viscometer holders. Bath temperature stabilizes within ±0.5°C (±1°F) of setting, and final adjustment to within ±0.1°C (±0.02°F) can be made. Test temperatures of up to 150°C (302°F) can be selected. Temperature limit control permits the operator to select an over-temperature cutoff point to protect against accidental overheating. Control unit includes immersion heater, circulatingMore Product Information