Three-Position and Six-Position Combination Heating Mantles, Glas-Col®
Supplier: Glas-Col
100DRX12512 100DRX12712 100DRX30412 100DRX30024 100DRX30424 100DRJ12512 100DRJ30412 100DRJ12524 100DRJ12724 100DRJ30024 100DRJ30424 100DRX50012 100DRX50212 100DRX65012 100DRX80012 100DRX100012 100DRX50024 100DRX50224 100DRX65024 100DRX80024 100DRX100024 100DRJ50012 100DRJ50212 100DRJ65012 100DRJ80012 100DRJ50024 100DRJ50224 100DRJ65024 100DRJ80024 100DRJ100024 104ARL3312 104ARL3324 104ARL3624 104ARL4312 104ARL4324 104ARL4624 100DRX12512E 100DRX12712E 100DRX30412E 100DRX30024E 100DRX30424E 100DRJ12512E 100DRJ12712E 100DRJ30012E 100DRJ30412E 100DRJ12524E 100DRJ12724E 100DRJ30024E 100DRJ30424E 100DRX50012E 100DRX50212E 100DRX65012E 100DRX80012E 100D RX100012E 100DRX50024E 100DRX50224E 100DRX65024E 100DRX80024E 100D RX100024E 100DRJ50012E 100DRJ50212E 100DRJ80012E 100DRJ50024E 100DRJ50224E 100DRJ65024E 100DRJ80024E 100D RJ100024E 100DRX30012 100DRJ30012 104ARL3612 104ARL4612 100DRX30012E
33795-250EA 4197.34 USD
33795-250 33795-252 33795-256 33795-258 33795-260 33795-262 33795-268 33795-270 33795-272 33795-274 33795-276 33795-278 33795-280 33795-282 33795-284 33795-286 33795-288 33795-290 33795-292 33795-294 33795-296 33795-298 33795-300 33795-302 33795-304 33795-306 33795-308 33795-310 33795-312 33795-314 33795-316 33795-318 33795-322 33795-324 33795-326 33795-330 33796-000 33796-004 33796-008 33796-010 33796-012 33796-014 33796-016 33796-018 33796-020 33796-022 33796-024 33796-026 33796-028 33796-030 33796-032 33796-034 33796-036 33796-038 33796-040 33796-042 33796-044 33796-046 33796-048 33796-050 33796-052 33796-054 33796-056 33796-058 33796-060 33796-062 33796-064 33795-254 33795-266 33795-320 33795-328 33796-006
Three-Position and Six-Position Combination Heating Mantles, Glas-Col®
Heaters Heating Mantles
Ideal for repetitive extracting, refluxing and distilling laboratory applications.
Low-profile combination mantle utilizes convenient heating element container system that allows quick and easy replacement of a failed element and eliminates the need to tear down entire system of glassware. Element containers also act as spill containment chambers. If spill occurs, the whole container can be economically replaced without damage to other parts of the mantle.
All models feature clear anodized aluminum cabinet with black fluoropolymer resin-coated stainless steel top. Three-position unitsMore Product Information