Casework, Laminate, Sitting Height Base Cabinets, Cupboard Cabinet
Supplier: Case Systems
B0110302424
33501-082EA 709.75 USD
33501-082
Casework, Laminate, Sitting Height Base Cabinets, Cupboard Cabinet
Furniture Casework
Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
  • Plastic laminate provides superior resistance to chemicals, scratching, and abrasion
  • The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards
  • The unique hardware system provides rigid support for vertical integration without limiting adjustability
The key component of case systems casework is the plasticMore Product Information