CircuitWorks® Flux Remover Pens, ITW Chemtronics®
Supplier: ITW Chemtronics
CW9200
33500-474EA 32.68 USD
33500-474
CircuitWorks® Flux Remover Pens, ITW Chemtronics®
Soldering Supplies Flux Removers
These fast-drying pens completely remove flux from printed circuit boards in seconds
Ideal for spot cleaning tight tolerance areas. Marker dispensers provide controlled, exact application.