KIMTECH PURE® G5 Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
56858
14215-736CS 790.56 USD
14215-736 32933-752 32933-758 32933-760 14214-532 14215-732 32933-754 32933-756 14215-734 14215-738
KIMTECH PURE® G5 Nitrile Gloves, Kimberly-Clark
Gloves
Ambidextrous, powder-free nitrile gloves contain no latex.
  • Static dissipative in use
  • Beaded cuff for easier donning
Light blue gloves have a bisque finish. White gloves have textured fingertips to facilitate control and grip.
Certifications: Recommended for ISO Class 5 (FED-STD-209E Class 100/M3.5) or higher cleanroom environments. Packaged in a Class 100 Cleanroom. AQL 1.5.
Ordering information: Packaged in a Class 100 cleanroom. Packed 100 per double poly bag with case liner.