PFA Autosampler Vials, Savillex
Supplier: SAVILLEX
200-002-44
30617-502EA 29.44 USD
30617-502
PFA Autosampler Vials, Savillex
Vials
PFA autosampler vials fit many GFAA autosamplers and ICP-MS micro autosamplers.
Service temperature range is –200 to 260°C (–328 to 500°F).
Vials feature a conical interior bottom for efficient sampling of microvolume samples. Manufactured from the highest purity PFA resin for ultratrace metals analysis. Snap-on PFA closure is sold separately.
Ordering information: Closure is not included, and must be ordered separately.