Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Supplier: Thermo Fisher Scientific
6320-0010
30049-122CS 285.86 USD
30049-122
Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Forceps
All-plastic forceps with double-action hinges for extra leverage and long life
Scissors-type handles with ratchet for locking tips shut. Interlocking teeth for tight grip. Provide a useful clamp for chromatography columns. Durable blue polypropylene is autoclavable for repeated use. Ideal for economical, disposable applications.