Hose Nuts, Western Enterprises
Supplier: Safety & Industrial Supplies
312-10 312-7 312-C-7 312-C-8 312-8 312-9 312-D-7 312-D-8
300007-780EA 0.98 USD
300007-780 300007-888 300007-953 300007-954 300067-028 300007-901 300007-907 300066-770 300066-772
Hose Nuts, Western Enterprises
Welding Supplies
  • Made from brass to ensure functionality and durability
  • Can accommodate up to 200 psi of pressure