Hose Couplers, Western Enterprises
Supplier: Safety & Industrial Supplies
312-130 312-30 312-C-31 312-D-31 312-231 312-D-30 312-31 312-C-30
300007-790EA 2.87 USD
300007-790 300007-819 300007-825 300066-688 300066-760 300066-302 300066-758 300007-791 300007-831 300007-950
Hose Couplers, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability.
Utilize hose couplers as a connector on the end of a hose to connect it with another hose or with a tap or a hose appliance.