Whatman™ Grade 113 Qualitative Filter Papers
Supplier: Cytiva
1113-125 1113-150 1113-185 1113-320 1113-500 1113-090 1113-110 1113-240 1113-917 1113-8768
28297-258PK 52.42 USD
28297-258 28297-260 28297-262 28464-980 28464-990 28297-254 10147-906 10147-908 10147-910 76423-772
Whatman™ Grade 113 Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous precipitates.
  • Fastest flow rate of the qualitative grades
  • Creped surface
  • Thickest filter paper in the qualitative range
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc. SomeMore Product Information