LABOPORT™ UN842 Vacuum Diaphragm Pumps, KNF
Supplier: KNF Neuberger Inc.
057634/042382 209649/047708
14224-136EA 9246.19 USD
14224-136 26678-020
LABOPORT™ UN842 Vacuum Diaphragm Pumps, KNF
Pumps Vacuum Diaphragm Pumps
Chemically resistant, oil-free diaphragm vacuum pump used for a wide range of laboratory applications. Especially suited for handling aggressive vapors, it features PTFE heads, PTFE-coated diaphragms and FFPM valves.
  • Handle corrosive and aggressive vapors without damage
  • 100% oil-free transfer
  • Highly compatible with vapours and condensation
  • Chemically resistant
  • Suitable for highly corrosive gases and vapours
  • High level of gas tightness
Pure transferring and evacuation of gases.
Order Now
SPECIFICATIONS
Environmentally Preferable
Flow Rate 34 L/min
Vacuum 1.5 torr
Protection class IP 44 enclosure rating
W×D×H 330.2×171.5×222.3 mm