ERGOMAT® Complete Smooth Anti-Fatigue Floor Mats, ERGOMAT
Supplier: Ergomat
SX0204 SX0214 SX0219 SX0226 SX0306 SX0316 SX0318 SX0319 SX0320 SX0321 SX0322 SX0330 SX0405 SX0307 SX0406 SX0206 SX0212 SX0215 SX0217 SX0220 SX0224 SX0229 SX0304 SX0310 SX0312 SX0313 SX0317 SX0323 SX0325 SX0407 SX0408 SX0411 SX0413 SX0414 SX0418 SX0419 SX0421 SX0426 SX0427 SX0429 SX0203 SX0207 SX0208 SX0210 SX0211 SX0213 SX0222 SX0225 SX0228 SX0230 SX0305 SX0308 SX0309 SX0326 SX0327 SX0328 SX0416 SX0417 SX0420 SX0423 SX0424 SX0425 SX0205 SX0209 SX0218 SX0221 SX0260 SX0303 SX0311 SX0314 SX0324 SX0409 SX0410 SX0412 SX0422 SX0428 SX0430 SX0216 SX0223 SX0227 SX0315 SX0329 SX0360 SX0404 SX0415 SX0460
22230-830EA 251.2 USD
22230-830 22230-850 22230-860 22230-874 22230-892 22230-912 22230-916 22230-918 22230-920 22230-922 22230-924 22230-940 22230-946 22230-894 22230-949 22230-834 22230-846 22230-852 22230-856 22230-862 22230-870 22230-880 22230-888 22230-900 22230-904 22230-906 22230-914 22230-926 22230-930 22230-950 22230-952 22230-958 22230-962 22230-964 22230-972 22230-974 22230-978 22230-988 22230-990 22230-994 22230-828 22230-836 22230-838 22230-842 22230-844 22230-848 22230-866 22230-872 22230-878 22230-882 22230-890 22230-896 22230-898 22230-932 22230-934 22230-936 22230-968 22230-970 22230-976 22230-982 22230-984 22230-986 22230-832 22230-840 22230-858 22230-864 22230-884 22230-886 22230-902 22230-908 22230-928 22230-954 22230-956 22230-960 22230-980 22230-992 22230-996 22230-854 22230-868 22230-876 22230-910 22230-938 22230-942 22230-944 22230-966 22230-998
ERGOMAT® Complete Smooth Anti-Fatigue Floor Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT Complete Smooth is a fire retardant, open cell, virgin polyurethane mat
  • Fire retardant, open cell, virgin polyurethane
  • Tolerates mild exposures of oil and other liquids
  • Silicone free, latex free, anti-static, UV resistant
  • Certified for use in ISO Class 5 (100) cleanrooms
It is designed to accommodate standing workers as well as cart and trolley traffic. It can also tolerate mild exposures of oil and other liquids. This silicone and latex free mat is anti-static, UV resistant and certified for use in ISO Class 5 (100) cleanrooms.