VertiKlean® Wall Washing Systems
Supplier: Contec
LVK-MOPIR MVK-MOPIR MVK-QT-SOX LVK-SOX TLVKMOP TMVKMOPIR TMVKMOP TLVKMOPIR TVKMOP VK-MOPIR 2746 LVK-MOP MVK-MOP VK-MOP TMVKMOP
12777-966CS 1083.68 USD
12777-966 12777-968 22221-806 33500-040 33501-058 33501-060 33501-062 33501-068 33501-072 68310-336 89068-854 89074-018 89198-836 68310-139 68310-143 68310-408 76239-976 76239-978
VertiKlean® Wall Washing Systems
Mops
Lightweight, polyester disposable mops are excellent for applying disinfectants and for wet cleaning of vertical surfaces, ceilings, and walls in controlled environments. Ideal for reaching awkward corners and ceilings with a complete range of motion.
  • Hydrophilic polyester fabric laminated to clean ester foam, laser cut sealed edge
  • Compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
  • Available in three sizes (8, 13 and 15")
For aseptic processing or sterile core areas, the sterile mop heads provide truly validatedMore Product Information