VWR® Talon® Nester Extension Clamp
Supplier: VWR International
21572-002
21572-002EA 151.23 USD
21572-002
VWR® Talon® Nester Extension Clamp
Clamps General Purpose Clamps
Securely holds 50–102 mm (2–4") diameter large or small glass distillation columns and odd-shaped glassware.
  • Stainless steel construction
  • Chromed-brass band and chain
  • Holds a variety of odd-shaped glassware
Constructed of stainless steel with strong, chromed-brass band and chain. Extension arm is 152 mm (6") long.