VWR® Talon® Three-Prong Extension Clamps
Supplier: VWR International
12621-236 97035-502 97035-504 12621-238 12621-240 21570-125 21570-126 21570-127 21570-200 21570-302 21570-404
12621-236EA 153.59 USD
12621-236 97035-502 97035-504 12621-238 12621-240 21570-125 21570-126 21570-127 21570-200 21570-302 21570-404
VWR® Talon® Three-Prong Extension Clamps
Clamps General Purpose Clamps
Clamps securely hold every type of glassware and apparatus.
  • Large grip adjustment range
  • Three-prong construction
  • Choose single or dual adjustment
  • Stainless steel or nickel-plated zinc
Long, seamless tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms that allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment. Stainless steel clamps offer exceptionalMore Product Information