Nalgene® Conical Oak Ridge Centrifuge Tube, Graduated, PPCO, Thermo Scientific
Supplier: Thermo Fisher Scientific
3148-0050
21010-615CS 1495.8 USD
21010-615
Nalgene® Conical Oak Ridge Centrifuge Tube, Graduated, PPCO, Thermo Scientific
Tubes Centrifuge Tubes
These conical centrifuge tubes combine the better separation/pelleting capabilities of a conical tube with the superior sealing capability of Oak Ridge tubes.
  • Graduations are silkscreened from 5 to 35 ml in 1 ml increments
The tubes accommodate Nalgene sealing caps and require conical tube adapter. The tubes are translucent but contact clear for easy liquid viewing.