VWR® High Performance Microcentrifuge Tubes, PP
Supplier: VWR International
20170-022 20170-024 20170-026 20170-028 20170-030 20170-032 20170-034 20170-036 20170-038
20170-022CS 534.75 USD
20170-022 20170-024 20170-026 20170-028 20170-030 20170-032 20170-034 20170-036 20170-038
VWR® High Performance Microcentrifuge Tubes, PP
Tubes Microcentrifuge Tubes
These tubes are made from USP Class VI PP, which ensures a clear view of the pellet. They incorporate a high-strength wall and can be centrifuged to twice the g-force that most other microcentrifuge tubes can withstand. Amber colored for light-sensitive substances.
  • Ideal for vortexing and shaking tables
  • RNase-, DNase-, and endotoxin-free
  • Engraved graduations ensure accuracy
  • Provide easy and legible mark area
  • Fit all common rotors
  • Ideal for phenol/chloroform extractions
The tubes also feature flat, pierceable, frosted caps and a side labeling surface.More Product Information