Iodine solution_standard 0.05 mol/l (0.1 N) water

Supplier: Thermo Scientific Chemicals
Warning

AC12422-0010
200057-888EA 97.03 USD
200057-888 103467-252
Iodine solution_standard 0.05 mol/l (0.1 N) water
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 1.02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3264
Merck Index: 13,05036

Order Now

Specification Test Results

Appearance Dark brown Solution
Titration with Na2S2O3 0.0998 to 0.1002 N (20 °C)
CO2 absorption capacity ≥19.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR