Nalgene® Packaging Bottles, HDPE, Narrow-Mouth, with Screw Caps, Thermo Scientific
Supplier: Thermo Fisher Scientific
312085-0001 312085-0002 312085-0004 312085-0008 312085-0016 312085-0032 312089-0001 312089-0002 312089-0004 312089-0008 312089-0016 312089-0032
16059-512CS 797.76 USD
16059-512 16059-514 16059-516 16059-518 16059-520 16059-522 16059-548 16059-550 16059-552 16059-554 16059-556 16059-558
Nalgene® Packaging Bottles, HDPE, Narrow-Mouth, with Screw Caps, Thermo Scientific
Bottles Plastic Bottles
These economical, break-resistant bottles are made from high-density polyethylene.
  • Available in natural or amber colors
  • Guaranteed leakproof
Natural sample bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Ordering information: Supplied with polypropylene screw caps.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification ISO14001 (Certificate Registration No. EMS 752603)