Temperature Controlled Shipper, Qorpak®
Supplier: Berlin Packaging
HMS-69110
15712-240CS 487.46 USD
15712-240
Temperature Controlled Shipper, Qorpak®
Mailers and Shippers
Certified packaging for shipment of category A infectious substances.
  • A leakproof, 95 kPa primary receptacle
  • A leakproof secondary receptacle
  • A strong outer container that has a minimum of one rigid side and 4” width
  • Packages acceptable for ground and air shipments
Certifications: UN Rated: 4GU / Class 6.2.
Delivery information: Kits come complete with absorbent, components and labels you need to be in compliance, with the exception of your primary receptacle.
Caution: Note: Sufficient absorbent material must be placed between the primary and secondary receptacles.