Stainless Steel Probe, Desco Industries
Supplier: Desco
35617 35630
15431-790EA 7.44 USD
15431-790 15431-796 15431-806 15431-808
Stainless Steel Probe, Desco Industries
Temperature Probes
The 5.5" stainless steel probe is ideal for working in ESD protective areas.
  • Ergonomic hex handle
  • Non-magnetic, conductive 300 series stainless steel
  • Stainless steel won’t corrode - no problem in working with magnetized parts
  • Any charge can be grounded when placed on ESD protected work surface
Made in the United States of America superior quality.
Delivery information: The stainless steel probe kit includes one each of the straight, hooked, angled, and curved probe versions.