Stainless Steel Worktables, Eagle MHC™
Supplier: Eagle Group
ET2424B ET2430B ET2436B ET2448B ET2460B ET2472B ET2496B ET3030B ET3036B ET3048B ET3060B ET3072B ET3096B
14900-800EA 672.64 USD
14900-800 14900-802 14900-804 14900-806 14900-808 14900-810 14900-812 14900-814 14900-816 14900-818 14900-820 14900-822 14900-824
Stainless Steel Worktables, Eagle MHC™
Furniture Tables and Workbenches
Worktables feature a 20-gauge, Type 430, polished stainless steel top, an adjustable 18-gauge galvanized steel undershelf, ext. Ø of 41 mm (¹³/₈") tubular heavy gauge galvanized steel legs, and 25 mm (1") adjustable high-impact plastic feet.
  • Unique uni-lok™ gusset system is recessed into the channel to reduce lateral movement
  • Undershelf is reinforced with a gusset welded to each corner
  • All tables are prepunched including attachment hardware for installation of drawer assemblies
Tabletop has 38 mm (³/₂") square edges that are flattened 180° at frontMore Product Information